missing translation for 'onlineSavingsMsg'
Learn More

Ethyl all cis-5,8,11,14,17-Eicosapentaenoate 96.0+%, TCI America™

Catalog No. E0442200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0442200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E0442200MG Supplier TCI America Supplier No. E0442200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86227-47-6
Molecular Formula C22H34O2
Molecular Weight (g/mol) 330.512
MDL Number MFCD00673476
InChI Key SSQPWTVBQMWLSZ-AAQCHOMXSA-N
Synonym all cis-5,8,11,14,17-Eicosapentaenoic Acid Ethyl Ester, EPA Ethyl Ester, Ethyl all cis-5,8,11,14,17-Icosapentaenoate
PubChem CID 9831415
ChEBI CHEBI:84883
IUPAC Name ethyl (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoate
SMILES CCC=CCC=CCC=CCC=CCC=CCCCC(=O)OCC

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 330.51
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl all cis-5,8,11,14,17-Eicosapentaenoate
RTECSNumber JX3840000
TSCA No
Recommended Storage Refrigerator