missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acrylate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. p-7116736
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A014325ML 25 mL
A0143500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A014325ML Supplier TCI America Supplier No. A014325ML

Chemical Identifiers

CAS 140-88-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009188
InChI Key JIGUQPWFLRLWPJ-UHFFFAOYSA-N
Synonym ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat
PubChem CID 8821
ChEBI CHEBI:82327
IUPAC Name ethyl prop-2-enoate
SMILES CCOC(=O)C=C

Specifications

Melting Point -71°C
Color Colorless
Boiling Point 100°C
Quantity 25 mL
UN Number 1917
Formula Weight 100.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0988
RTECSNumber AT0700000
TSCA Yes
Recommended Storage Refrigerator