missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetoacetate 98.0+%, TCI America™

Catalog No. p-7116985
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
A064925G 25 g
A0649500G 500 g
2 options

Catalog No. A064925G

Supplier: TCI America A064925G

Only null left

Chemical Identifiers

CAS 141-97-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00009199
InChI Key XYIBRDXRRQCHLP-UHFFFAOYSA-N
Synonym ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
PubChem CID 8868
ChEBI CHEBI:4893
IUPAC Name ethyl 3-oxobutanoate
SMILES CCOC(=O)CC(=O)C

Specifications

Melting Point -43°C
Color Colorless
Boiling Point 184°C
Quantity 25 g
Formula Weight 130.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Acetoacetate
EINECSNumber (2)-1475&(2)-1505
RTECSNumber AK5250000
TSCA Yes