missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, Technical, Spectrum™ Chemical
Specifications
Specifications
| Quantity | 500 mL |
| Packaging | Steel Can |
| Molecular Formula | C4H8O2 |
| Boiling Point | 77°C |
| InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
| SMILES | CCOC(C)=O |
| IUPAC Name | ethyl acetate |
| Grade | Technical |
| Chemical Name or Material | Ethyl Acetate |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.