missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, Technical, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 4 L |
| Packaging | Steel Can |
| Boiling Point | 77°C |
| Grade | Technical |
| Chemical Name or Material | Ethyl Acetate |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.