missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate for Spectroscopy Uvasol™ MilliporeSigma™

Catalog No. M1008630500
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1008630500 500 mL Glass Bottle
M1008632500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1008630500 Supplier Technidata Supplier No. 1.00863.0500

For spectroscopy Uvasol™

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83°C
Color Colorless
Density 0.90g/cm3 at 20°C
Quantity 500 mL
Flash Point −4°C (−25°F)
Ignition Point 460°C (860°F)
Odor Fruity
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.37
Vapor Pressure 97 hPa (20°C)
UN Number 1173
Boiling Point 77°C
Solubility Information Soluble in water: 85.3g/L at 20°C
Formula Weight 88.11
Chemical Name or Material Ethyl Acetate
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H319: Causes serious eye irritation.
Precautionary Statement H336: May cause drowsiness or dizziness.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.