missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC), Solstice

Catalog No. 60046222
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
180 kg
Packaging:
Glass Bottle
Metal Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046222 2.5 L Glass Bottle
60046221 1 L Glass Bottle
60046226 6 x 1 L Glass Bottle
60046225 4 x 2.5 L Glass Bottle
60046220 180 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60046222 Supplier Solstice Supplier No. 332112.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Density 0.902g/mL at 25°C
Quantity 2.5 L
Explosion Limit 2.2 to 11.5% (38°F)
Flash Point −3°C (26.6°F)
Ignition Point 801°F
Odor Like fruit
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 73mmHg (20°C)
Linear Formula CH3COOC2H5
UN Number UN1173
Beilstein 506104
Boiling Point 77°C
Solubility Information ∽90g/L
Formula Weight 88.11g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/ISO/Reagent Ph. Eur./puriss. p.a.
Chemical Name or Material Ethyl acetate
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
DOTInformation Class 3 - PG 2 - Ethyl acetate
RTECSNumber 205-500-4