missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, puriss., ≥99.5% (GC); Solstice

Catalog No. 60045769
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
35 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
180 kg
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Poly Bottle
Stainless Steel Can
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045769 6 x 1 L Poly Bottle
60045763 1 L Glass Bottle
60045762 1 L Poly Bottle
60045764 2.5 L Poly Bottle
60045768 5 L Aluminum Bottle
60045766 4 x 2.5 L Poly Bottle
60045767 4 x 5 L Aluminum Bottle
60045765 35 kg Stainless Steel Can
60045761 180 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 60045769 Supplier Solstice Supplier No. 272276X1L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Density 0.898 to 0.902g/mL at 20°C, 0.894 to 0.898g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 2.2 to 11.5% (38°F)
Flash Point −3°C (26.6°F)
Ignition Point 801°F
Packaging Poly Bottle
Physical Form Liquid
Linear Formula CH3COOC2H5
Vapor Pressure 73mmHg (20°C)
UN Number UN1173
Beilstein 506104
Boiling Point 76.5°C to 77.5°C
Formula Weight 88.11g/mol
Percent Purity ≥99.5% (GC)
Grade puriss.
Chemical Name or Material Ethyl Acetate, Meets analytical specification of Ph. Eur.BP, NF
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H319-H336
DOTInformation Class 3 - PG 2 - Ethyl acetate
EINECSNumber 205-500-4
RTECSNumber AH5425000