missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, puriss., ≥99.5% (GC); Honeywell™ Riedel-de Haën™
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | -84°C |
| Color | Colorless |
| Density | 0.898 to 0.902g/mL at 20°C, 0.894 to 0.898g/mL at 25°C |
| Quantity | 5 L |
| Explosion Limit | 2.2 to 11.5% (38°F) |
| Flash Point | −3°C (26.6°F) |
| Ignition Point | 801°F |
| Packaging | Aluminum Bottle |
| Physical Form | Liquid |
| Vapor Pressure | 73mmHg (20°C) |
| Show More |
Safety and Handling
| Hazard Statement | P210-P305 + P351 + P338-P370 + P378-P403 + P235 |
| Precautionary Statement | H225-H319-H336 |
| DOTInformation | Class 3 - PG 2 - Ethyl acetate |
| EINECSNumber | 205-500-4 |
| RTECSNumber | AH5425000 |