missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, puriss., ≥99.5% (GC); Honeywell™ Riedel-de Haën™
Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99.5% (GC)
Supplier: Honeywell-Riedel-de Haen 2722735KG
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
| Ethyl Acetate | |
| Meets analytical specification of Ph. Eur.BP, NF | |
| Colorless | |
| 76.5°C to 77.5°C | |
| 2.2 to 11.5% (38°F) | |
| 801°F | |
| Liquid | |
| CH3COOC2H5 | |
| 73mmHg (20°C) | |
| 506104 | |
| XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
| ethyl acetate | |
| 8857 | |
| 88.11g/mol | |
| puriss. |
| -84°C | |
| 141-78-6 | |
| 0.898 to 0.902g/mL at 20°C, 0.894 to 0.898g/mL at 25°C | |
| 35 kg | |
| −3°C (26.6°F) | |
| Stainless Steel Can | |
| C4H8O2 | |
| MFCD00009171 | |
| UN1173 | |
| ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
| CCOC(=O)C | |
| 88.106 | |
| CHEBI:27750 | |
| ≥99.5% (GC) |
Chemical Identifiers
| 141-78-6 | |
| 88.106 | |
| XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
| 8857 | |
| ethyl acetate |
| C4H8O2 | |
| MFCD00009171 | |
| ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
| CHEBI:27750 | |
| CCOC(=O)C |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
H225-H319-H336
DOTInformation : Class 3 - PG 2 - Ethyl acetate
EINECSNumber : 205-500-4
RTECSNumber : AH5425000