missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, puriss., ≥99.5% (GC); Honeywell™ Riedel-de Haën™

Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99.5% (GC)

Supplier:  Honeywell-Riedel-de Haen 272271L

 View more versions of this product

Catalog No. 60045762


Only null left
Explore available promotions

Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Specifications

Ethyl Acetate
Meets analytical specification of Ph. Eur.BP, NF
Colorless
76.5°C to 77.5°C
2.2 to 11.5% (38°F)
801°F
Liquid
CH3COOC2H5
73mmHg (20°C)
506104
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11g/mol
puriss.
-84°C
141-78-6
0.898 to 0.902g/mL at 20°C, 0.894 to 0.898g/mL at 25°C
1 L
−3°C (26.6°F)
Poly Bottle
C4H8O2
MFCD00009171
UN1173
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CCOC(=O)C
88.106
CHEBI:27750
≥99.5% (GC)

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P305 + P351 + P338-P370 + P378-P403 + P235

H225-H319-H336

DOTInformation : Class 3 - PG 2 - Ethyl acetate

EINECSNumber : 205-500-4

RTECSNumber : AH5425000

SDS
Documents

Documents

Promotions

Promotions