missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.5%, Solstice

Catalog No. 6000595
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000595 19 L Metal Can
6000596 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000595 Supplier Solstice Supplier No. PP10019

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

CAS Max % 100.0000%
Density 0.8945 g/cm3
Quantity 19 L
Flash Point −4°C
Packaging Metal Can
UN Number UN1173
Water 0.1% max.
Beilstein 506104
Boiling Point 77°C
Solubility Information 870% in Water at 20°C
Formula Weight 88.11
Percent Purity >99.9%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Ethyl Acetate, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate