missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, Optima™ for HPLC and GC, Thermo Scientific™

Catalog No. p-215931
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Safe-Cote™ Amber Glass 
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
E1964 4 L Amber Soda Lime Glass
E196SK4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1964 Supplier Thermo Fisher Scientific Supplier No. FLE1964
Only null left
Edge

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.5°C
Absorbance 0.015 AU max. at 270nm, 0.12 AU max. at 260nm, 1.00 AU max. at 255nm
Color Colorless
Density 0.902g/cm³
Quantity 4 L
Identification Pass Test
Packaging Amber Soda Lime Glass
Physical Form Liquid
Refractive Index 1.3680 to 1.3710 (at 25°C)
Residue after Evaporation 1ppm max.
Assay Percent Range Assay: 99.9% min by GC
Titratable Acid 0.0009mEq/g max.
Linear Formula CH3COOC2H5
Vapor Pressure 103mbar at 20°C
Viscosity 0.45 cP at 20°C
Water 0.02% max.
Boiling Point 75°C
Formula Weight 88.11
Percent Purity ≥99.9%
Grade High purity mobile phase for HPLC and/or extraction solvent for GC applications
Chemical Name or Material Ethyl Acetate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid