missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, OmniSolv™, MilliporeSigma™

Catalog No. MEX02411
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Poly-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MEX02411 4 L Glass Bottle
MEX0241P1 4 L Poly-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MEX02411 Supplier Technidata Supplier No. EX02411

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For environmental analysis

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Melting Point -84°C
Color Colorless
Density 0.90g/cm3 at 20°C
Quantity 4 L
Flash Point -4°C (-25°F)
Ignition Point 425.85°C (798.5°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
UN Number UN 1173
Solubility Information Partially soluble in water
Boiling Point 77.2°C
Formula Weight 88.12
Percent Purity ≥99.9%
Chemical Name or Material Ethyl Acetate
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.