missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, LC-MS CHROMASOLV™, Honeywell Riedel-de Haën™

Catalog No. p-7072147
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000578 1 L
6000579 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000578 Supplier Honeywell Chemicals Supplier No. 349721L
Only null left

CHROMASOLV™ LC-MS

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

 

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Density 0.902g/mL at 25°C
Quantity 1 L
Explosion Limit 2.2 to 11.5%, 38°F
Flash Point −3°C (26.6°F)
Ignition Point 801°F
Linear Formula CH3COOC2H5
Vapor Pressure 73 mmHg (20°C)
UN Number UN1173
Beilstein 506104
Formula Weight 88.11g/mol
Percent Purity ≥99.7% (GC)
Grade LC-MS CHROMASOLV
Chemical Name or Material Ethyl acetate
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
RTECSNumber AH5425000