missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate (HPLC), Fisher Chemical™

Catalog No. p-215936
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
E1954 4 L Glass Bottle
E195SK4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1954 Supplier Thermo Fisher Scientific Supplier No. E195 4
Only null left
Edge

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.5°C
Absorbance 1 AU max. at 255nm, 0.15 AU max. at 260nm, 0.025 AU max. at 270nm
Color Colorless
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range Assay: 99.9% min by GC
Titratable Acid 0.0009mEq/g max.
Linear Formula CH3COOC2H5
Vapor Pressure 103mbar at 20°C
Viscosity 0.45 cP at 20°C
UN Number 1173
Water 0.02% max.
Boiling Point 75°C
Formula Weight 88.11
Percent Purity ≥99.9%
Grade Mobile phase for HPLC applications
Chemical Name or Material Ethyl Acetate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid