missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, Exceeds A.C.S. Specifications, HPLC Grade, 99.8%, Spectrum™ Chemical

Catalog No. 18610396
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610396 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18610396 Supplier Spectrum Chemical Mfg Cor Supplier No. HP6024LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141-78-6
Molecular Weight (g/mol) 88.11

Specifications

CAS Max % 1
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 3 ppm
Titratable Acid 0.0009 meq/g
Water 0.0005
Boiling Point 77°C
Percent Purity 0.998
Grade ACS/HPLC
Chemical Name or Material Ethyl Acetate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.