missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, Environmental Grade, 99.5+%

Catalog No. AA40977K2
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40977K2 1 L
AA40977M1 4 x 1 L
AA40977K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40977K2 Supplier Thermo Scientific Chemicals Supplier No. 040977K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pesticide residue, environmental, and GC analysis | CAS: 141-78-6 | C4H8O2 | 88.106 g/mol

Ethyl acetate is used primarily as a solvent and diluent. It is commonly used to clean the circuit boards, in some nail varnish removers and also in residue, environmental and GC analysis. It is used chiefly as a scent in the manufacture of perfumes. It is also used in flavorings, glues, in decaffeinating tea and coffee, and in cigarettes. The other minor applications of ethyl acetate include its applications in the textiles, dyestuffs, explosives, photographic films and plates, artificial leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Melting Point -84°C
Color Colorless
Density 0.902 g/mL
Quantity 1 L
Flash Point -3°C (26°F)
Odor Sweet
Physical Form Liquid
Refractive Index 1.372
Linear Formula CH3CO2CH2CH3
UN Number UN1173
Beilstein 506104
Merck Index 14,3757
Boiling Point 77°C
Solubility Information Miscible with ethanol,acetone,diethyl ether and benzene.
Formula Weight 88.11
Percent Purity ≥99.5%
Grade Environmental
Chemical Name or Material Ethyl acetate
Show More Show Less
Hazard Category H225-H319-H336
Hazard Statement GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL ACETATE
EINECSNumber 205-500-4
RTECSNumber AH5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only