missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, MilliporeSigma™

Catalog No. MEX02456
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MEX02456 1 L Glass Bottle
MEX02451 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MEX02456 Supplier Technidata Supplier No. EX02456

For HPLC

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Density 0.90g/cm3 (20°C)
Quantity 1 L
Explosion Limit 2.1 to 11.5%(V)
Packaging Glass Bottle
Vapor Pressure 97 hPa (20°C)
Linear Formula CH3COOC2H5
Formula Weight 88.11
Chemical Name or Material Ethyl Acetate
Recommended Storage Store at +15°C to +25°C