missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate CHROMASOLV™, for HPLC, ≥99.7%, Solstice

Catalog No. 6000574
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000574 4 x 2.5 L
6000569 1 L
6000570 2.5 L
6000575 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Repeated exposure may cause skin dryness or cracking.
Catalog No. 6000574 Supplier Solstice Supplier No. 348584X2.5L
Repeated exposure may cause skin dryness or cracking.

CHROMASOLV™, for HPLC,≥99.7%

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Density 0.902g/mL at 25°C
Quantity 4 x 2.5 L
Refractive Index n20/D 1.3720
Vapor Pressure 73mmHg (20°C)
Linear Formula CH3COOC2H5
Beilstein 506104
Boiling Point 76.5°C to 77.5°C
Formula Weight 88.11g/mol
Percent Purity ≥99.7%
Grade CHROMASOLV
Chemical Name or Material Ethyl acetate, for HPLC
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
Precautionary Statement H225-H319-H336
EINECSNumber 205-500-4
RTECSNumber AH5425000