missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl acetate CHROMASOLV™, for HPLC, ≥99.7%, Solstice
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -84°C |
| Density | 0.902g/mL at 25°C |
| Quantity | 2.5 L |
| Refractive Index | n20/D 1.3720 |
| Vapor Pressure | 73mmHg (20°C) |
| Linear Formula | CH3COOC2H5 |
| Beilstein | 506104 |
| Boiling Point | 76.5°C to 77.5°C |
| Formula Weight | 88.11g/mol |
| Percent Purity | ≥99.7% |
| Show More |
Safety and Handling
| Hazard Statement | P210-P261-P305 + P351 + P338 |
| Precautionary Statement | H225-H319-H336 |
| EINECSNumber | 205-500-4 |
| RTECSNumber | AH5425000 |