missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, For pesticide residue analysis, Solstice

Catalog No. 60045892
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60045892 4 x 2.5 L
6000580 1 L
6000581 2.5 L
6000582 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60045892 Supplier Solstice Supplier No. 310634X2.5L

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 0.902 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Flash Point −4°C
Packaging Glass Bottle
Linear Formula CH3COOC2H5
UN Number UN1173
Beilstein 506104
Boiling Point 77°C
Formula Weight 88.11
Percent Purity >99.9%
Grade Pesticide
Chemical Name or Material Ethyl acetate, For pesticide residue analysis
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate
EINECSNumber 205-500-4
Recommended Storage Room Temp