missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023045
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023045 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023045 Supplier Merck Emd Millipore Supplier No. RTC00008720ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These secondary standards offer multi-traceability to the USP, EP, and BP Primary Standards, where they are available.

Specifications

Melting Point -84°C (literature)
CAS 141-78-6
Density 0.902 g/mL (at 25°C (literature))
Quantity 20 mL
Flash Point -3°C (Not Applicable)
Refractive Index n20/D 1.372 (literature)
Molecular Formula C4H8O2
Vapor Pressure 73 mmHg (20°C)
Linear Formula CH3COOC2H5
MDL Number MFCD00009171
Synonym Ethyl acetate; EtOAc
Boiling Point 76.5°C to 77.5°C (literature)
Formula Weight 88.11
Grade Reference Material
Show More Show Less