missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, B&J Brand™, for HPLC, for spectrophotometry, >99.5%, Honeywell Burdick & Jackson

Catalog No. p-7112023
Change view
Click to view available options
Quantity:
56 L
200 L
Packaging:
Returnable Steel Drum
Catalog No. Quantity Packaging
6000587 56 L Returnable Steel Drum
6000583 200 L Returnable Steel Drum
2 options

Catalog No. 6000587

Supplier: Honeywell Chemicals CS10056

Only null left

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Absorbance 0.05 AU max. at 275 nm, 0.03 AU max. at 300 nm, 0.005 AU max. at 325 nm, 0.005 AU max. at 350 nm
CAS Max % 100.0000%
Density 0.8945 g/cm3
Boiling Point 77°C
Quantity 56 L
Flash Point −4°C
Packaging Returnable Steel Drum
UN Number UN1173
Beilstein 506104
Water 0.03% max.
Solubility Information 870% in Water at 20°C
Formula Weight 88.11
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Ethyl Acetate, B&J Brand™, for HPLC, for spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate