missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, ≥99.9%, Honeywell Burdick & Jackson

Catalog No. p-7111739
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Catalog No. Quantity Packaging
6000588 6 x 1 L Glass Bottle
6000586 4 x 4 L Glass Bottle
2 options

Catalog No. 6000588

Supplier: Honeywell Chemicals 1001L

Only null left

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Absorbance 0.05 AU max. at 275 nm (UV), 0.03 AU max. at 300 nm (UV), 0.005 AU max. at 325 nm (UV), 0.005 AU max. at 400 nm (UV)
CAS Max % 100.0000%
Density 0.8945 g/cm3
Boiling Point 77°C
Quantity 6 x 1 L
Flash Point −4°C
Packaging Glass Bottle
UN Number UN1173
Beilstein 506104
Formula Weight 88.11
Percent Purity ≥99.9%
Grade HPLC
Chemical Name or Material Ethyl Acetate, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, ≥99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate