missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, ≥99.9%, Honeywell Burdick & Jackson
Specifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.8945 g/cm3 |
| Quantity | 20 L |
| Flash Point | −4°C |
| Packaging | HDPE Nowpak™ |
| UN Number | UN1173 |
| Beilstein | 506104 |
| Boiling Point | 77°C |
| Formula Weight | 88.11 |
| Percent Purity | >99.5% |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate |