missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, ACS Reagent, Reagents

Catalog No. 01337992
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337992 1 L
01337991 20 L
01337990 4 L
01337989 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337992 Supplier Labchem Inc Supplier No. C11995001C
Only null left

Ethyl Acetate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym Ethyl Ethanoate, Acetic Acid Ethyl Ester, Acetic Ether
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Melting Point -83.6°C
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.8946
Boiling Point 77°C
Grade ACS
Chemical Name or Material Ethyl Acetate, ACS Reagent