missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Honeywell

Catalog No. p-7112044
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
Packaging:
Glass Bottle
Metal Can
Poly Bottle
Catalog No. Quantity Packaging
6000593 4 x 4 L Glass Bottle
6000594 4 x 4 L Poly Bottle
6000591 19 L Metal Can
3 options

Catalog No. 6000593

Supplier: Honeywell Chemicals LP1004

Only null left

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

CAS Max % 100.0000%
Density 0.8945 g/cm3
Boiling Point 77°C
Quantity 4 x 4 L
Flash Point −4°C
Packaging Glass Bottle
UN Number UN1173
Beilstein 506104
Water 0.2% max.
Solubility Information 870% in Water at 20°C
Formula Weight 88.11
Percent Purity >99.5%
Grade ACS Reagent
Assay by GC Analysis
Chemical Name or Material Ethyl Acetate, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate