missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, ≥99.5%, Honeywell™

Catalog No. p-7112004
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000589 4 x 4 L Glass Bottle
6000590 4 x 4 L Poly-coated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 6000589 missing translation for 'mfr' Honeywell Chemicals missing translation for 'supplierNo' AH1004
Only null left

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

CAS Max % 100.0000%
Density 0.8945 g/cm3
Boiling Point 77°C
Quantity 4 x 4 L
Flash Point −4°C
Packaging Glass Bottle
UN Number UN1173
Beilstein 506104
Water 0.03% max.
Solubility Information 870% in Water at 20°C
Formula Weight 88.11
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material Ethyl Acetate, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, ≥99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl acetate
EINECSNumber 205-500-4