missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate ACS, MilliporeSigma™

Catalog No. p-3176630
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Metal Can
Poly-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MEX0240P4 4 L Poly-coated Glass Bottle
MEX02403 20 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MEX0240P4 Supplier Technidata Supplier No. EX0240P4
Only null left

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Boiling Point 77.2°C
Quantity 4 L
Flash Point -4°C (-25°F)
Ignition Point 425.85°C (798.5°F)
Odor Charactristic
Packaging Poly-coated Glass Bottle
Physical Form Liquid
Linear Formula CH3COOCH2CH3
UN Number UN 1173
Solubility Information Partially soluble in water
Formula Weight 88.11
Percent Purity ≥99.5%
Grade ACS Reagent
Chemical Name or Material Ethyl Acetate
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II