missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, 99.5% min., ACS, MilliporeSigma™
Supplier: Technidata EX024020
Specifications
| Ethyl Acetate | |
| 141-78-6 | |
| 200 L | |
| 99.5% min. | |
| ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
| CCOC(=O)C | |
| 88.106 | |
| CHEBI:27750 |
| For ACS analysis | |
| Colorless | |
| Liquid | |
| C4H8O2 | |
| XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
| ethyl acetate | |
| 8857 | |
| 88.11 |
Chemical Identifiers
| 141-78-6 | |
| 88.106 | |
| ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
| CHEBI:27750 | |
| CCOC(=O)C |
| C4H8O2 | |
| XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
| 8857 | |
| ethyl acetate |