missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, 99.5% min., ACS, MilliporeSigma™

Catalog No. MEX02405
Change view
Click to view available options
Quantity:
4 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MEX02405 4 L
MEX024020 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MEX02405 Supplier Technidata Supplier No. US1EX0240/5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Color Colorless
Quantity 4 L
Physical Form Liquid
Assay Percent Range 99.5% min.
Formula Weight 88.11
Chemical Name or Material Ethyl Acetate, For ACS analysis