missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, ≥99.5%, ACS Reagent, Solstice

Catalog No. 60046011
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
6 x 1 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046011 500 mL Glass Bottle
60046013 6 x 500 mL Glass Bottle
60046012 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046011 Supplier Solstice Supplier No. 319902500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Density 0.902g/mL at 25°C
Quantity 500 mL
Explosion Limit 2.2 to 11.5% (38°F)
Flash Point −3°C (26.6°F)
Ignition Point 801°F
Packaging Glass Bottle
Linear Formula CH3COOC2H5
Vapor Pressure 73mmHg (20°C)
UN Number UN1173
Beilstein 506104
Boiling Point 77.5°C
Formula Weight 88.11g/mol
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Ethyl Acetate
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
Precautionary Statement H225-H319-H336
DOTInformation Class 3 - PG 2 - Ethyl acetate
EINECSNumber 205-500-4
RTECSNumber AH5425000