missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, Laboratory Reagent, ≥99.0% (GC), Solstice

Catalog No. 60044939
Change view
Click to view available options
Quantity:
4 x 5 L
Packaging:
Aluminum Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044939 4 x 5 L Aluminum Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60044939 Supplier Solstice Supplier No. 163714X5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory Reagent,≥99.0% (GC)

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Density 0.902g/mL at 25°C
Quantity 4 x 5 L
Explosion Limit 2.2 to 11.5% (38°F)
Flash Point −3°C (26.6°F)
Ignition Point 801°F
Packaging Aluminum Bottle
Vapor Pressure 73mmHg (20°C)
Linear Formula CH3COOC2H5
UN Number UN1173
Beilstein 506104
Formula Weight 88.11g/mol
Percent Purity ≥99.0% (GC)
Grade Laboratory
Chemical Name or Material Ethyl Acetate
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H319-H336
DOTInformation class 3 - PG 2 - Ethyl acetate
EINECSNumber 205-500-4
RTECSNumber AH5425000