missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, EMPLURA™, MilliporeSigma™

Catalog No. 361042000
Change view
Click to view available options
Quantity:
5 L
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361042000 5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361042000 Supplier Technidata Supplier No. 8.22277.5000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83°C
Density 0.90g/cm3
Quantity 5 L
Explosion Limit 2.1 - 11.5%(V)
Flash Point -4°C
Ignition Point 460°C
Packaging Plastic Bottle
Residue after Evaporation ≤0.003%
Assay Percent Range ≥99.5% (a/a)
Linear Formula C4H8O2
Vapor Pressure 97 hPa
UN Number UN 1173
Boiling Point 777 deg;C (at 1013 hPa)
Formula Weight 88.11
Chemical Name or Material Ethyl Acetate
Show More Show Less
Hazard Category Danger
Hazard Statement H225: Highly flammable liquid and vapour. H319: Causes serious eye irritation. H336: May cause drowsiness or dizziness. EUH066: Repeated exposure may cause skin dryness or cracking.
Precautionary Statement P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking. P240: Ground/bond container and receiving equipment. P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P403 + P233: Store in a well-ventilated place. Keep container tightly closed.
EINECSNumber 205-500-4
RTECSNumber AH5425000
Recommended Storage Store at +2°C to +25°C