missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, 97%, Thermo Scientific™

Catalog No. BTB02003EB
Change view
Click to view available options
Quantity:
1g
10g
25g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB02003EB 25g Amber Glass Bottle
BTB02003DA 1g Amber Glass Bottle
BTB02003EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB02003EB Supplier Thermo Scientific Chemicals Supplier No. BTB02003EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71083-06-2
Molecular Formula C12H10FNO3
Molecular Weight (g/mol) 235.214
MDL Number MFCD00052242
InChI Key MPUYCZQHTGRPNE-UHFFFAOYSA-N
Synonym ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate, ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate, 3-quinolinecarboxylicacid, 8-fluoro-1,4-dihydro-4-oxo-, ethyl ester, 8-fluoro-4-hydroxy-quinoline-3-carboxylic acid ethyl ester, 8-fluoro-4-hydroxyquinoline-3-carboxylic acid ethyl ester, 3-quinolinecarboxylic acid, 8-fluoro-4-hydroxy-, ethyl ester, pubchem19599, maybridge1_000540, acmc-209nc3
PubChem CID 707160
IUPAC Name ethyl 8-fluoro-4-oxo-1H-quinoline-3-carboxylate
SMILES CCOC(=O)C1=CNC2=C(C1=O)C=CC=C2F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25g
Formula Weight 235.21
Percent Purity 97%
Chemical Name or Material Ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.