missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB02003EA

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. BTB02003EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

71083-06-2
235.214
MPUYCZQHTGRPNE-UHFFFAOYSA-N
707160
CCOC(=O)C1=CNC2=C(C1=O)C=CC=C2F
C12H10FNO3
MFCD00052242
ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate, ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate, 3-quinolinecarboxylicacid, 8-fluoro-1,4-dihydro-4-oxo-, ethyl ester, 8-fluoro-4-hydroxy-quinoline-3-carboxylic acid ethyl ester, 8-fluoro-4-hydroxyquinoline-3-carboxylic acid ethyl ester, 3-quinolinecarboxylic acid, 8-fluoro-4-hydroxy-, ethyl ester, pubchem19599, maybridge1_000540, acmc-209nc3
ethyl 8-fluoro-4-oxo-1H-quinoline-3-carboxylate

Specifications

Ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate
96.5
97%
C12H10FNO3
MFCD00052242
MPUYCZQHTGRPNE-UHFFFAOYSA-N
ethyl 8-fluoro-4-oxo-1H-quinoline-3-carboxylate
707160
97%
71083-06-2
100.0
Amber Glass Bottle
10g
ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate, ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate, 3-quinolinecarboxylicacid, 8-fluoro-1,4-dihydro-4-oxo-, ethyl ester, 8-fluoro-4-hydroxy-quinoline-3-carboxylic acid ethyl ester, 8-fluoro-4-hydroxyquinoline-3-carboxylic acid ethyl ester, 3-quinolinecarboxylic acid, 8-fluoro-4-hydroxy-, ethyl ester, pubchem19599, maybridge1_000540, acmc-209nc3
CCOC(=O)C1=CNC2=C(C1=O)C=CC=C2F
235.214
235.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents