missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate, 95%

Catalog No. AAH3591603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3591603 1 g
AAH3591606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3591603 Supplier Thermo Scientific Chemicals Supplier No. H3591603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 353258-31-8 | C11H8ClF3N2O2 | 292.642 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 353258-31-8
Molecular Formula C11H8ClF3N2O2
Molecular Weight (g/mol) 292.642
MDL Number MFCD01833013
InChI Key AKACORAIGDTIFR-UHFFFAOYSA-N
Synonym ethyl 8-chloro-6-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylate, 8-chloro-6-trifluoromethyl-imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, ethyl 8-chloro-6-trifluoromethyl imidazo 1,2-a-pyridine-2-carboxylate, 4,6-dimethylbenzofuran-3-carboxylic acid, ethyl 8-chloro-6-trifluoromethyl-imidazo 1,2-a pyridine-2-carboxylate, 8-chloro-6-trifluoromethyl imidazo 1,2,a pyridine-2-carboxylic acid ethyl ester, 8-chloro-6-trifluoromethyl-imidazo1,2-apyridine-2-carboxylic acid ethyl ester
PubChem CID 737427
IUPAC Name ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
SMILES CCOC(=O)C1=CN2C=C(C=C(C2=N1)Cl)C(F)(F)F

Specifications

Melting Point 132°C to 136°C
Quantity 1 g
Formula Weight 292.65
Percent Purity 95%
Chemical Name or Material Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only