missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%
CAS: 50551-58-1 | C12H12O4 | 220.22 g/mol
Supplier: Thermo Scientific Chemicals B2106606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl 7-methoxybenzo[b]furan-2-carboxylate | |
| 83°C to 86°C | |
| MFCD00060510 | |
| ethyl 7-methoxybenzofuran-2-carboxylate, 7-methoxybenzofuran-2-carboxylic acid ethyl ester, 7-methoxybenzofuran-2-carboxylic acid, ethyl ester, ethyl 7-methoxybenzo b furan-2-carboxylate, 7-methoxybenzofuran-2-carboxylic acidethyl ester, ethyl7-methoxybenzofuran-2-carboxylate, acmc-1aowz, 2-ethoxycarbonyl-7-methoxybenzofuran, ethyl 4-methoxylbenzofunan-2-carboxylate, ethyl 7-methoxybenzo d furan-2-carboxylate | |
| CCOC(=O)C1=CC2=CC=CC(OC)=C2O1 | |
| 220.22 | |
| 220.22 |
| 50551-58-1 | |
| C12H12O4 | |
| 5 g | |
| SXCMZYVXJUJIDJ-UHFFFAOYSA-N | |
| ethyl 7-methoxy-1-benzofuran-2-carboxylate | |
| 521284 | |
| 97% |
Chemical Identifiers
| 50551-58-1 | |
| 220.22 | |
| SXCMZYVXJUJIDJ-UHFFFAOYSA-N | |
| ethyl 7-methoxy-1-benzofuran-2-carboxylate |
| C12H12O4 | |
| MFCD00060510 | |
| 521284 | |
| CCOC(=O)C1=CC2=CC=CC(OC)=C2O1 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only