missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-azaindole-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAH3425003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3425003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3425003 Supplier Thermo Scientific Chemicals Supplier No. H3425003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1261588-72-0
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.202
MDL Number MFCD15071133
InChI Key CKCBQXQNAUHBDX-UHFFFAOYSA-N
PubChem CID 56773892
IUPAC Name ethyl 1H-pyrrolo[2,3-b]pyridine-4-carboxylate
SMILES CCOC(=O)C1=C2C=CNC2=NC=C1

Specifications

Melting Point 127°C to 129°C
Quantity 1 g
Synonym ethyl 1h-pyrrolo 2,3-b pyridine-4-carboxylate, c9e, ethylpyrrolobpyridinecarboxylate, ethyl 7-azaindole-4-carboxylate, ethyl1h-pyrrolo 2,3-b pyridine-4-carboxylate, 1h-pyrrolo 2,3-b pyridine-4-carboxylic acid, ethyl ester
Formula Weight 190.2
Percent Purity 97%
Chemical Name or Material Ethyl 7-azaindole-4-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only