missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-Hydroxyhexanoate 95.0+%, TCI America™

Catalog No. E13055G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E13055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E13055G Supplier TCI America Supplier No. E13055G
Only null left

Chemical Identifiers

CAS 5299-60-5
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.21
MDL Number MFCD00075601
InChI Key HYXRUZUPCFVWAH-UHFFFAOYSA-N
Synonym 6-Hydroxyhexanoic Acid Ethyl Ester
PubChem CID 357781
IUPAC Name ethyl 6-hydroxyhexanoate
SMILES CCOC(=O)CCCCCO

Specifications

Color Colorless
Boiling Point 128°C
Quantity 5 g
Formula Weight 160.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 6-Hydroxyhexanoate