missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-chloroindole-2-carboxylate, 97%

Catalog No. p-7057122
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6648303 1 g
AAH6648306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6648303 Supplier Thermo Scientific Chemicals Supplier No. H6648303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27034-51-1 | C11H10ClNO2 | 223.656 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27034-51-1
Molecular Formula C11H10ClNO2
Molecular Weight (g/mol) 223.656
MDL Number MFCD03084731
InChI Key FSMZLIBWSAMADK-UHFFFAOYSA-N
Synonym ethyl 6-chloroindole-2-carboxylate, 6-chloroindole-2-carboxylic acid ethyl ester, 6-chloro-1h-indole-2-carboxylic acid ethyl ester, 6-chloro-2-indolecarboxylic methyl ester, 1h-indole-2-carboxylic acid, 6-chloro-, ethyl ester, pubchem1672, ethyl 6-chloroindole-2 carboxylate, ethyl-6-chloroindole-2-carboxylate, ethyl 6-chloranyl-1h-indole-2-carboxylate, 6-chlorindole-2-carboxylic acid ethyl ester
PubChem CID 7015027
IUPAC Name ethyl 6-chloro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=C(C=C2)Cl

Specifications

Quantity 1 g
Beilstein 169178
Formula Weight 223.66
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 6-chloroindole-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only