missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-(Chloroformyl)hexanoate 97.0+%, TCI America™

Catalog No. C14145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C14145G Supplier TCI America Supplier No. C14145G

Chemical Identifiers

CAS 14794-32-2
Molecular Formula C9H15ClO3
Molecular Weight (g/mol) 206.666
MDL Number MFCD00671507
InChI Key NYZNYLOIPHDFKS-UHFFFAOYSA-N
Synonym ethyl 6-chloroformyl hexanoate, 6-chloroformyl hexanoic acid ethyl ester, heptanoic acid,7-chloro-7-oxo-, ethyl ester, acmc-209czj, ethyl 6-chloroformylhexanoate, ethyl 6-chloroformyl-hexanoate, 6-chlorocarbonylhexanoic acid ethyl ester, heptanoic acid, 7-chloro-7-oxo, ethyl ester
PubChem CID 543232
IUPAC Name ethyl 7-chloro-7-oxoheptanoate
SMILES CCOC(=O)CCCCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 141°C
Quantity 5 g
UN Number 3265
Formula Weight 206.67
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl 6-(Chloroformyl)hexanoate