missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 6-Bromopyridine-2-carboxylate 98.0+%, TCI America™
Supplier: TCI America E08265G
Specifications
| Ethyl 6-Bromopyridine-2-carboxylate | |
| 41°C | |
| C8H8BrNO2 | |
| 5 g | |
| OONBOXSIWCBAHZ-UHFFFAOYSA-N | |
| ethyl 6-bromopyridine-2-carboxylate | |
| 2758810 | |
| ≥98.0% (GC) |
| 21190-88-5 | |
| White-Yellow | |
| MFCD04116913 | |
| 6-Bromopicolinic Acid Ethyl Ester, Ethyl 6-Bromopicolinate, 6-Bromopyridine-2-carboxylic Acid Ethyl Ester | |
| CCOC(=O)C1=NC(=CC=C1)Br | |
| 230.061 | |
| 230.06 | |
| Crystalline Powder |
Chemical Identifiers
| 21190-88-5 | |
| 230.061 | |
| OONBOXSIWCBAHZ-UHFFFAOYSA-N | |
| 2758810 | |
| CCOC(=O)C1=NC(=CC=C1)Br |
| C8H8BrNO2 | |
| MFCD04116913 | |
| 6-Bromopicolinic Acid Ethyl Ester, Ethyl 6-Bromopicolinate, 6-Bromopyridine-2-carboxylic Acid Ethyl Ester | |
| ethyl 6-bromopyridine-2-carboxylate |
Safety and Handling
TSCA : No