missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate, 98%

Catalog No. p-7032508
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3399203 1 g
AAH3399206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3399203 Supplier Thermo Scientific Chemicals Supplier No. H3399203
Only null left

CAS: 1027511-41-6 | C9H8BrN3O2 | 270.086 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1027511-41-6
Molecular Formula C9H8BrN3O2
Molecular Weight (g/mol) 270.086
MDL Number MFCD10574908
InChI Key PZZTXIWJXOMKEU-UHFFFAOYSA-N
Synonym ethyl 6-bromopyrazolo 1,5-a pyrimidine-3-carboxylate, 6-bromo-3-ethoxycarbonyl pyrazolo 1,5-a pyrimidine, 6-bromopyrazolo 1,5-a pyrimidine-3-carboxylic acid ethyl ester, pubchem20934, ethyl 6-bromopyrazolo 1,5-a pyrimidine-3-carboxyla, ethyl 6-bromo-pyrazolo 1,5-a pyrimidine-3-carboxylate, ethyl 6-bromopyrazolo 1,5-alpha pyrimidine-3-carboxylate, 6-bromo-pyrazolo 1,5-a pyrimidine-3-carboxylic acid ethyl ester
PubChem CID 45073325
IUPAC Name ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate
SMILES CCOC(=O)C1=C2N=CC(=CN2N=C1)Br

Specifications

Melting Point 144°C to 146°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 270.08
Percent Purity 98%
Chemical Name or Material Ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only