missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-Bromocoumarin-3-carboxylate 98.0+%, TCI America™

Catalog No. p-7129120
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07335G 5 g
E073325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07335G Supplier TCI America Supplier No. E07335G
Only null left

Chemical Identifiers

CAS 2199-90-8
Molecular Formula C12H9BrO4
Molecular Weight (g/mol) 297.104
MDL Number MFCD00124584
InChI Key RXGDNKCWEKUYQA-UHFFFAOYSA-N
Synonym 6-Bromocoumarin-3-carboxylic Acid Ethyl Ester
PubChem CID 691747
IUPAC Name ethyl 6-bromo-2-oxochromene-3-carboxylate
SMILES CCOC(=O)C1=CC2=C(C=CC(=C2)Br)OC1=O

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 5 g
Formula Weight 297.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 6-Bromocoumarin-3-carboxylate