missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]hexanoate 95.0+%, TCI America™

Catalog No. p-7129431
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1313100MG 100 mg
E1313500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1313100MG Supplier TCI America Supplier No. E1313100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 572913-59-8
Molecular Formula C29H38NO6P
Synonym 6-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]hexanoic Acid Ethyl Ester

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 527.60
Percent Purity ≥95.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Ethyl 6-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]hexanoate
TSCA No
Recommended Storage Refrigerator