missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 5-hydroxyindole-2-carboxylate, 98+%
CAS: 24985-85-1 | C11H11NO3 | 205.213 g/mol
$156.75 - $524.30
Chemical Identifiers
| CAS | 24985-85-1 |
|---|---|
| Molecular Formula | C11H11NO3 |
| Molecular Weight (g/mol) | 205.213 |
| MDL Number | MFCD00015458 |
| InChI Key | WANAXLMRGYGCPC-UHFFFAOYSA-N |
| Synonym | ethyl 5-hydroxyindole-2-carboxylate, 5-hydroxy-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-hydroxy-, ethyl ester, 5-hydroxyindole-2-carboxylic acid ethyl ester, ethyl-5-hydroxy-1h-indol-2-carboxylat, pubchem15850, acmc-1cs3a, 5-hydroxy-1h-indole-2-carboxylicacidethylester, 2-carbethoxy-5-hydroxy-indole, ethyl 5-hydroxy-1h-2-indolecarboxylate |
| PubChem CID | 90677 |
| IUPAC Name | ethyl 5-hydroxy-1H-indole-2-carboxylate |
| SMILES | CCOC(=O)C1=CC2=C(N1)C=CC(=C2)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0077403
|
Thermo Scientific Chemicals
L0077403 |
1 g |
Each for $156.75
|
|
|||||
|
AAL0077406
|
Thermo Scientific Chemicals
L0077406 |
5 g |
Each for $524.30
|
|
|||||
Description
Ethyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store between 2-8 C. Store in cool, dryplace in a well ventilated area. Store in a tightly sealed container. Protect from moisture, heat.
Chemical Identifiers
| 24985-85-1 | |
| 205.213 | |
| WANAXLMRGYGCPC-UHFFFAOYSA-N | |
| 90677 | |
| CCOC(=O)C1=CC2=C(N1)C=CC(=C2)O |
| C11H11NO3 | |
| MFCD00015458 | |
| ethyl 5-hydroxyindole-2-carboxylate, 5-hydroxy-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-hydroxy-, ethyl ester, 5-hydroxyindole-2-carboxylic acid ethyl ester, ethyl-5-hydroxy-1h-indol-2-carboxylat, pubchem15850, acmc-1cs3a, 5-hydroxy-1h-indole-2-carboxylicacidethylester, 2-carbethoxy-5-hydroxy-indole, ethyl 5-hydroxy-1h-2-indolecarboxylate | |
| ethyl 5-hydroxy-1H-indole-2-carboxylate |
Specifications
| 24985-85-1 | |
| C11H11NO3 | |
| 1 g | |
| Light sensitive | |
| Insoluble in water. | |
| CCOC(=O)C1=CC2=C(N1)C=CC(=C2)O | |
| 205.213 | |
| 205.21 | |
| Ethyl 5-hydroxyindole-2-carboxylate |
| 150°C to 151°C | |
| MFCD00015458 | |
| 476925 | |
| ethyl 5-hydroxyindole-2-carboxylate, 5-hydroxy-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-hydroxy-, ethyl ester, 5-hydroxyindole-2-carboxylic acid ethyl ester, ethyl-5-hydroxy-1h-indol-2-carboxylat, pubchem15850, acmc-1cs3a, 5-hydroxy-1h-indole-2-carboxylicacidethylester, 2-carbethoxy-5-hydroxy-indole, ethyl 5-hydroxy-1h-2-indolecarboxylate | |
| WANAXLMRGYGCPC-UHFFFAOYSA-N | |
| ethyl 5-hydroxy-1H-indole-2-carboxylate | |
| 90677 | |
| ≥98% |
Safety and Handling
EINECSNumber : 246-554-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only