missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 5-hydroxyindole-2-carboxylate, 98+%
Description
Ethyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store between 2-8 C. Store in cool, dryplace in a well ventilated area. Store in a tightly sealed container. Protect from moisture, heat.
Ethyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store between 2-8 C. Store in cool, dryplace in a well ventilated area. Store in a tightly sealed container. Protect from moisture, heat.
Specifications
Specifications
| Melting Point | 150°C to 151°C |
| Quantity | 1 g |
| Beilstein | 476925 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 205.21 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Ethyl 5-hydroxyindole-2-carboxylate |
Safety and Handling
| EINECSNumber | 246-554-9 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only