missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Formyl-2,4-dimethyl-3-pyrrolecarboxylate 98.0+%, TCI America™

Catalog No. E10515G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10515G 5 g
E10511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10515G Supplier TCI America Supplier No. E10515G
Only null left

Chemical Identifiers

CAS 2199-59-9
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00030352
InChI Key GDISALBEIGGPER-UHFFFAOYSA-N
Synonym 5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylic acid ethyl ester, 2,4-dimethyl-3-ethoxycarbonyl-5-formylpyrrole, ethyl 2,4-dimethyl-5-formylpyrrole-3-carboxylate, ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl-, ethyl ester, ethyl 3,5-dimethyl-2-formylpyrrole-4-carboxylate, 2,4-dimethyl-3-carbethoxy pyrrole-5-carboxaldehyde, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrrole-2-carboxaldehyde, 5-formyl-2,4-dimethyl-3-pyrrolecarboxylic acid ethyl ester, pubchem9415
PubChem CID 137485
IUPAC Name ethyl 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
SMILES CCOC(=O)C1=C(C)NC(C=O)=C1C

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 5-Formyl-2,4-dimethyl-3-pyrrolecarboxylate